Anti-KRT19 Mouse Monoclonal Antibody [clone: A53-B/A2.26]

Proveedor: OriGene
ORIGTA327664EA 813 EUR
ORIGTA327664
Anti-KRT19 Mouse Monoclonal Antibody [clone: A53-B/A2.26]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: A53-B/A2.26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a lambda
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR