Anti-KRT14 Mouse Monoclonal Antibody [clone: LL002]

Proveedor: OriGene
ORIGTA327662EA 746 EUR
ORIGTA327662
Anti-KRT14 Mouse Monoclonal Antibody [clone: LL002]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: KRT14
Clonality: Monoclonal
Clone: LL002
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR