Anti-CEACAM5 Mouse Monoclonal Antibody

Proveedor: OriGene
ORIGTA327659EA 622 EUR
ORIGTA327659
Anti-CEACAM5 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: CEACAM5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR