Anti-CEACAM5 Mouse Monoclonal Antibody [clone: CEA31]

Proveedor: OriGene
ORIGTA327658EA 466 EUR
ORIGTA327658
Anti-CEACAM5 Mouse Monoclonal Antibody [clone: CEA31]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: CEACAM5
Clonality: Monoclonal
Clone: CEA31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR