Anti-CD63 Mouse Monoclonal Antibody [clone: NKI/C3]

Proveedor: OriGene
ORIGTA327649EA 1020 EUR
ORIGTA327649
Anti-CD63 Mouse Monoclonal Antibody [clone: NKI/C3]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: CD63
Clonality: Monoclonal
Clone: NKI/C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR