Anti-B3GAT1 Mouse Monoclonal Antibody [clone: NK1]

Proveedor: OriGene
ORIGTA327647EA 677 EUR
ORIGTA327647
Anti-B3GAT1 Mouse Monoclonal Antibody [clone: NK1]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: B3GAT1
Clonality: Monoclonal
Clone: NK1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR