Anti-SLC2A3 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA325844EA 450 EUR
ORIGTA325844
Anti-SLC2A3 Rabbit Polyclonal Antibody
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: SLC2A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR