Anti-GRIN1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326171EA 564 EUR
ORIGTA326171
Anti-GRIN1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:100 Immunofluorescence: 1:100-1:500 ELISA: 1:40000

Type: Primary
Antigen: GRIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR