Anti-TP53 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326166EA 564 EUR
ORIGTA326166
Anti-TP53 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:100 ELISA: 1:10000Western Blot: 1:500-1:3000 Immunoprecipitation: Various Dilution

Type: Primary
Antigen: TP53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR