Anti-NCAM1 Mouse Monoclonal Antibody [clone: 123C3.D5]

Proveedor: OriGene
ORIGTA327645EA 814 EUR
ORIGTA327645
Anti-NCAM1 Mouse Monoclonal Antibody [clone: 123C3.D5]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: 123C3.D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR