Anti-CD33 Mouse Monoclonal Antibody [clone: PWS44]

Proveedor: OriGene
ORIGTA327636EA 1380 EUR
ORIGTA327636
Anti-CD33 Mouse Monoclonal Antibody [clone: PWS44]
Recommended Dilutions: Immunochemistry: 1:20-1:80

Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: PWS44
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR