Anti-CD3D Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA327632EA 895 EUR
ORIGTA327632
Anti-CD3D Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:1000

Type: Primary
Antigen: CD3D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR