Anti-MS4A1 Mouse Monoclonal Antibody [clone: L26]

Proveedor: OriGene
ORIGTA327626EA 466 EUR
ORIGTA327626
Anti-MS4A1 Mouse Monoclonal Antibody [clone: L26]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: L26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR