Anti-CD2 Mouse Monoclonal Antibody [clone: MRQ-11]

Proveedor: OriGene
ORIGTA327625EA 895 EUR
ORIGTA327625
Anti-CD2 Mouse Monoclonal Antibody [clone: MRQ-11]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:10-1:50

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: MRQ-11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR