Anti-CD1A Mouse Monoclonal Antibody [clone: MTB1]

Proveedor: OriGene
ORIGTA327623EA 1300 EUR
ORIGTA327623
Anti-CD1A Mouse Monoclonal Antibody [clone: MTB1]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:10-1:50

Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: MTB1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR