Anti-CD14 Rabbit Monoclonal Antibody [clone: EPR3653]

Proveedor: OriGene
ORIGTA327620EA 1040 EUR
ORIGTA327620
Anti-CD14 Rabbit Monoclonal Antibody [clone: EPR3653]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: EPR3653
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR