Anti-SDC1 Mouse Monoclonal Antibody [clone: B-A38]

Proveedor: OriGene
ORIGTA327619EA 922 EUR
ORIGTA327619
Anti-SDC1 Mouse Monoclonal Antibody [clone: B-A38]
Recommended Dilutions: Immunochemistry: 1:25-1:200

Type: Primary
Antigen: SDC1
Clonality: Monoclonal
Clone: B-A38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR