Anti-KIT Rabbit Monoclonal Antibody [clone: YR145]

Proveedor: OriGene
ORIGTA327618EA 922 EUR
ORIGTA327618
Anti-KIT Rabbit Monoclonal Antibody [clone: YR145]
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: KIT
Clonality: Monoclonal
Clone: YR145
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR