Anti-ITGAX Mouse Monoclonal Antibody [clone: 5D11]

Proveedor: OriGene
ORIGTA327617EA 1180 EUR
ORIGTA327617
Anti-ITGAX Mouse Monoclonal Antibody [clone: 5D11]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:20-1:80

Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: 5D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR