Anti-MME Mouse Monoclonal Antibody [clone: 56C6]

Proveedor: OriGene
ORIGTA327616EA 1680 EUR
ORIGTA327616
Anti-MME Mouse Monoclonal Antibody [clone: 56C6]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:10-1:50

Type: Primary
Antigen: MME
Clonality: Monoclonal
Clone: 56C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR