Anti-CNN1 Mouse Monoclonal Antibody [clone: CALP]

Proveedor: OriGene
ORIGTA327613EA 979 EUR
ORIGTA327613
Anti-CNN1 Mouse Monoclonal Antibody [clone: CALP]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: CNN1
Clonality: Monoclonal
Clone: CALP
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR