Anti-OAS1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326756EA 454 EUR
ORIGTA326756
Anti-OAS1 Goat Polyclonal Antibody
Recommended Dilutions: ELISA: 1:128000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: OAS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near C-terminus
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR