Anti-HOXB6 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326753EA 500 EUR
ORIGTA326753
Anti-HOXB6 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1-3?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: HOXB6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR