Anti-SIRT3 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326133EA 496 EUR
ORIGTA326133
Anti-SIRT3 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000 ELISA: 1:5000

Type: Primary
Antigen: SIRT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR