Anti-SHC3 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326108EA 514 EUR
ORIGTA326108
Anti-SHC3 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:100 ELISA: 1:10000

Type: Primary
Antigen: SHC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR