Anti-C3 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA327608EA 1180 EUR
ORIGTA327608
Anti-C3 Rabbit Polyclonal Antibody
Anticuerpos
  • Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: C3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR