Anti-BCL2 Rabbit Monoclonal Antibody [clone: E17]

Proveedor: OriGene
ORIGTA327607EA 924 EUR
ORIGTA327607
Anti-BCL2 Rabbit Monoclonal Antibody [clone: E17]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:25-1:100

Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: E17
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR