Anti-PRDX6 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326746EA 547 EUR
ORIGTA326746
Anti-PRDX6 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:128000; Western Blot: 0.1-0.3µg/ml

Type: Primary
Antigen: PRDX6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near N-terminus
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR