Anti-STX11 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326736EA 547 EUR
ORIGTA326736
Anti-STX11 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:128000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: STX11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR