Anti-JUND Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326732EA 550 EUR
ORIGTA326732
Anti-JUND Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:64000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: JUND
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR