Anti-NR5A2 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326727EA 547 EUR
ORIGTA326727
Anti-NR5A2 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:128000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: NR5A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR