Anti-APOD Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326725EA 502 EUR
ORIGTA326725
Anti-APOD Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 0.3-1?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: APOD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR