Anti-SERPINA1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326724EA 500 EUR
ORIGTA326724
Anti-SERPINA1 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:16000; Western Blot: 0.1-0.3µg/ml

Type: Primary
Antigen: SERPINA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR