Anti-SCG2 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326722EA 454 EUR
ORIGTA326722
Anti-SCG2 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:128000; Western Blot: 0.3-1µg/ml

Type: Primary
Antigen: SCG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR