Anti-CAV1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326097EA 466 EUR
ORIGTA326097
Anti-CAV1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000 Immunofluorescence: 1:100-1:500ELISA: 1:10000

Type: Primary
Antigen: CAV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR