Anti-FYN Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326081EA 466 EUR
ORIGTA326081
Anti-FYN Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000 ELISA: 1:1000

Type: Primary
Antigen: FYN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR