Anti-PAK1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326080EA 543 EUR
ORIGTA326080
Anti-PAK1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100 ELISA: 1:20000

Type: Primary
Antigen: PAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR