Anti-EIF4EBP1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326075EA 466 EUR
ORIGTA326075
Anti-EIF4EBP1 Rabbit Polyclonal Antibody
Anticuerpos
  • Recommended Dilutions: Immunochemistry: 1:50-1:100 ELISA: 1:40000

Type: Primary
Antigen: EIF4EBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR