Anti-MAP1LC3B Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326683EA 512 EUR
ORIGTA326683
Anti-MAP1LC3B Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: MAP1LC3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR