Anti-SUMO3 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA327516EA 563 EUR
ORIGTA327516
Anti-SUMO3 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-1:2000; Immunochemistry: 1:100-1:300

Type: Primary
Antigen: SUMO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR