Anti-AIMP2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326670EA 618 EUR
ORIGTA326670
Anti-AIMP2 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: AIMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR