Anti-IL36B Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326666EA 576 EUR
ORIGTA326666
Anti-IL36B Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: IL36B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR