Anti-ERAP1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326650EA 605 EUR
ORIGTA326650
Anti-ERAP1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1-2?g/ml

Type: Primary
Antigen: ERAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR