Anti-FLI1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326644EA 576 EUR
ORIGTA326644
Anti-FLI1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 0.5-1?g/ml

Type: Primary
Antigen: FLI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR