Anti-SARM1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326626EA 576 EUR
ORIGTA326626
Anti-SARM1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: SARM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR