Anti-ACP5 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326625EA 618 EUR
ORIGTA326625
Anti-ACP5 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: ACP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR