Anti-LMNA Rabbit Polyclonal Antibody [clone: Poly6135]

Proveedor: OriGene
ORIGTA328054EA 525 EUR
ORIGTA328054
Anti-LMNA Rabbit Polyclonal Antibody [clone: Poly6135]
Anticuerpos
Type: Primary
Antigen: LMNA
Clonality: Polyclonal
Clone: Poly6135
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, weakly cross-reactive with Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR