Anti-STAT1 Mouse Monoclonal Antibody [clone: SAT-84]

Proveedor: OriGene
ORIGTA328053EA 303 EUR
ORIGTA328053
Anti-STAT1 Mouse Monoclonal Antibody [clone: SAT-84]
Type: Primary
Antigen: STAT1
Clonality: Monoclonal
Clone: SAT-84
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR