Anti-MAPK1 Rabbit Polyclonal Antibody [clone: Poly6242]

Proveedor: OriGene
ORIGTA328051EA 333 EUR
ORIGTA328051
Anti-MAPK1 Rabbit Polyclonal Antibody [clone: Poly6242]
Anticuerpos
Type: Primary
Antigen: MAPK1
Clonality: Polyclonal
Clone: Poly6242
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR