Anti-BAX Mouse Monoclonal Antibody [clone: 6A7]

Proveedor: OriGene
ORIGTA328050EA 475 EUR
ORIGTA328050
Anti-BAX Mouse Monoclonal Antibody [clone: 6A7]
Anticuerpos
Type: Primary
Antigen: BAX
Clonality: Monoclonal
Clone: 6A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR